Learn More
U.S. Pharmacopeia Succinylcholine chloride, 6101-15-1, MFCD00150564, 500 mg
Catalog No. NC1447018
Encompass
Linear Formula: [(CH3)3NCH2CH2OCOCH2]2Cl2 · 2H2O, Molecular Weight: 397.34, Synonym: Bis(trimethylammonioethyl) succinate chloride, Succinylcholine chloride dihydrate, Succinyldicholine dichloride.
Catalog No. NC1447018 Supplier U.S. Pharmacopeia Supplier No. 1623502
Linear Formula: [(CH3)3NCH2CH2OCOCH2]2Cl2 · 2H2O, Molecular Weight: 397.34, Synonym: Bis(trimethylammonioethyl) succinate chloride, Succinylcholine chloride dihydrate, Succinyldicholine dichloride.