Learn More
TARGETMOL CHEMICALS INC Succinylcholine Chloride Dihyd
SDP
Catalog No. 502876904
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Succinylcholine Chloride Dihydrate (Succinyldicholine dichloride) is a nicotinic AChR agonist and also acts as a depolarizing neuromuscular blocker. Purity 99.91%
Catalog No. 50-287-6904 Supplier TARGETMOL CHEMICALS INC Supplier No. T6684100MG
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Succinylcholine Chloride Dihydrate (Succinyldicholine dichloride) is a nicotinic AChR agonist and also acts as a depolarizing neuromuscular blocker. Purity 99.91%