Learn More
Sigma Aldrich Fine Chemicals Biosciences Succinylcholine Chloride P1G
Catalog No. 502897293
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Catalog No. 50-289-7293 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR26591G
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.