Learn More
Medchemexpress LLC Succinylsulfathiazole | 116-43-8 | MFCD00022437 | 99.2% | 250 MG
SDP
Catalog No. 5000407371
Encompass_Preferred
Succinylsulfathiazole is a long-acting, orally active sulfonamide antibiotic with localized gut-specific antibacterial activity. It inhibits bacterial folate synthesis, leading to reduced coliform counts, suppressed intestinal bacterial growth, and depletion of gut folate-producing bacteria. Beyond its antibacterial role, it modulates hepatic mTOR signaling, diminishes cecal fermentation, and alters folate and nitrogen excretion. This compound can also induce folate deficiency and related nutritional deficiency symptoms.
  • Long-acting and orally active sulfonamide antibiotic
  • Provides localized gut-specific antibacterial activity
  • Inhibits bacterial folate synthesis
  • Reduces coliform counts and suppresses intestinal bacterial growth
  • Modulates hepatic mTOR signaling
  • Diminishes cecal fermentation
  • Decreases hepatic folate levels and total folate excretion
  • Can induce folate deficiency
Catalog No. 50-004-07371 Supplier Medchemexpress LLC Supplier No. 250MGHYB0921
Succinylsulfathiazole is a long-acting, orally active sulfonamide antibiotic with localized gut-specific antibacterial activity. It inhibits bacterial folate synthesis, leading to reduced coliform counts, suppressed intestinal bacterial growth, and depletion of gut folate-producing bacteria. Beyond its antibacterial role, it modulates hepatic mTOR signaling, diminishes cecal fermentation, and alters folate and nitrogen excretion. This compound can also induce folate deficiency and related nutritional deficiency symptoms.
  • Long-acting and orally active sulfonamide antibiotic
  • Provides localized gut-specific antibacterial activity
  • Inhibits bacterial folate synthesis
  • Reduces coliform counts and suppresses intestinal bacterial growth
  • Modulates hepatic mTOR signaling
  • Diminishes cecal fermentation
  • Decreases hepatic folate levels and total folate excretion
  • Can induce folate deficiency