Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinylsulfathiazole 97.0+%, TCI America™
SDP

Catalog No. S0991100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0991100G 100 g
S099125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0991100G Supplier TCI America Supplier No. S0991100G

Chemical Identifiers

CAS 116-43-8
Molecular Formula C13H13N3O5S2
Molecular Weight (g/mol) 355.383
MDL Number MFCD00022437
InChI Key SKVLYVHULOWXTD-UHFFFAOYSA-N
Synonym succinylsulfathiazole, cremosuxidine, sulfasuccidine, sulfasuccinil, sulfasuxidine, colistatin, sulfadigesin, sulfasuccithiazole, sulfasuccidin, sulfenterone
PubChem CID 5315
ChEBI CHEBI:9309
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES C1=CC(=CC=C1NC(=O)CCC(=O)O)S(=O)(=O)NC2=NC=CS2

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 100 g
Formula Weight 355.38
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Succinylsulfathiazole
RTECSNumber WM4767000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.