Learn More
Sigma Aldrich Fine Chemicals Biosciences Succnyl-beta-cyclodextrn500MG
Catalog No. 502900193
Encompass
Succinyl--cyclodextrin is a negatively charged cyclodextrin derivative that has major implications in the chiral separation of neutral and basic analytes by means of capillary electrophoresis (CE) owing to its counter-current mobility.
Catalog No. 50-290-0193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 85990500MG
Succinyl--cyclodextrin is a negatively charged cyclodextrin derivative that has major implications in the chiral separation of neutral and basic analytes by means of capillary electrophoresis (CE) owing to its counter-current mobility.