Learn More
Cayman Chemical Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA (hydrochloride), Purity: ≥95% (mixture of tautomers), CAS: 1379822-04-4, 10 mg
SDP
Catalog No. 502940986
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the IE(γ-pip)GR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-294-0986 Supplier Cayman Chemical Supplier No. 2770710 MG
A colorimetric Factor Xa substrate; cleavage of the IE(γ-pip)GR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity