Learn More
Cayman Chemical Succinimidyl 4-formylbenzoate, Purity: ≥95%, CAS: 60444-78-2, 10 mg
SDP
Catalog No. 502990834
Encompass_Preferred
A heterobifunctional building block; has been used in the conjugation of oligonucleotides to antibodies for protein detection, as well as in the synthesis of fluorescent BODIPY-etoposide
Catalog No. 50-299-0834 Supplier Cayman Chemical Supplier No. 3047710 MG
A heterobifunctional building block; has been used in the conjugation of oligonucleotides to antibodies for protein detection, as well as in the synthesis of fluorescent BODIPY-etoposide