Learn More
Aviva Systems Biology Corp SUCLA2 Antibody - N-terminal region (ARP64676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUCLA2. It was validated on Western Blot. . Alternate names: A-BETA, SCS-betaA, MTDPS5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-333