Learn More
Aviva Systems Biology Corp SUCLA2 Antibody - C-terminal region (ARP64675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUCLA2. It was validated on Western Blot. . Alternate names: A-BETA, SCS-betaA, MTDPS5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-332