Learn More
Cayman Chemical SucLeuLeuValTyrAMCtrIf 100mg
SDP
Catalog No. 502915302
Encompass_Preferred
A sesquiterpene lactone with anticancer activity inhibits the proliferation of MDA-MB-231 MDA-MB-468 and MCF-7 human breast cancer cells (IC50s 5.211 3.405 and 9.083 UM respectively after 48 hours) inhibits migration of MDA-MB-231 cells at 2.5 5 and 10 UM reduces tumor volume in an MDA-MB-231 orthotopic mouse xenograft model at 25 and 50 mg/kg
Catalog No. 50-291-5302 Supplier Cayman Chemical Supplier No. 37043100 MG
A sesquiterpene lactone with anticancer activity inhibits the proliferation of MDA-MB-231 MDA-MB-468 and MCF-7 human breast cancer cells (IC50s 5.211 3.405 and 9.083 UM respectively after 48 hours) inhibits migration of MDA-MB-231 cells at 2.5 5 and 10 UM reduces tumor volume in an MDA-MB-231 orthotopic mouse xenograft model at 25 and 50 mg/kg