Learn More
Cayman Chemical Suc-Leu-Leu-Val-Tyr-AMC (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502915262
Encompass_Preferred
A fluorogenic substrate for the 20S proteasome and calpains; AMC is released upon enzymatic cleavage by proteases and its fluorescence can be used to quantify 20S proteasome and calpain activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-291-5262 Supplier Cayman Chemical Supplier No. 3704350 MG
A fluorogenic substrate for the 20S proteasome and calpains; AMC is released upon enzymatic cleavage by proteases and its fluorescence can be used to quantify 20S proteasome and calpain activity; ex/em = 340-360/440-460 nm, respectively