Learn More
Aviva Systems Biology Corp SUCLG1 Antibody - C-terminal region (ARP65389_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65389_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUCLG1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human SUCLG1. Alternate names: FLJ21114, FLJ43513, GALPHA, SUCLA1, MTDPS9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-490