Learn More
Aviva Systems Biology Corp SUCLG1 Antibody - N-terminal region (ARP65390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUCLG1. It was validated on Western Blot. . Alternate names: FLJ21114, FLJ43513, GALPHA, SUCLA1, MTDPS9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-288-491