Learn More
Selleck Chemical LLC Sucralfate S4670-20mg
SDP
Catalog No. 503111000
Encompass_Preferred
Sucralfate(Sucrose octasulfate-aluminum complex) is a sucrose sulfate-aluminium complex that binds to the ulcer creating a physical barrier that protects the gastrointestinal tract from stomach acid and prevents the degradation of mucus
Catalog No. 50-311-1000 Supplier Selleck Chemical LLC Supplier No. S467020MG
Sucralfate(Sucrose octasulfate-aluminum complex) is a sucrose sulfate-aluminium complex that binds to the ulcer creating a physical barrier that protects the gastrointestinal tract from stomach acid and prevents the degradation of mucus