Learn More
Apexbio Technology LLC Sucralose 56038-13-2 50mg
SDP
Catalog No. 502240712
Encompass_Preferred
Sucralose (CAS 56038-13-2) is a synthetic chlorinated disaccharide widely employed as a noncaloric sweetener in biomedical and nutritional research It functions by binding to the sweet taste receptors (T1R2/T1R3) on the tongue eliciting a perceived sweetness substantially greater than that of sucrose Sucralose is minimally metabolized in the human body resulting in negligible caloric contribution and low systemic absorption Its metabolic stability and lack of glycemic impact make it a useful tool for investigating carbohydrate metabolism taste receptor signaling pathways and the physiological effects of nonnutritive sweeteners
This item is not returnable. View return policy
Catalog No. 50-224-0712 Supplier Apexbio Technology LLC Supplier No. B184050
Sucralose (CAS 56038-13-2) is a synthetic chlorinated disaccharide widely employed as a noncaloric sweetener in biomedical and nutritional research It functions by binding to the sweet taste receptors (T1R2/T1R3) on the tongue eliciting a perceived sweetness substantially greater than that of sucrose Sucralose is minimally metabolized in the human body resulting in negligible caloric contribution and low systemic absorption Its metabolic stability and lack of glycemic impact make it a useful tool for investigating carbohydrate metabolism taste receptor signaling pathways and the physiological effects of nonnutritive sweeteners