Learn More
Medchemexpress LLC Sucralose | 56038-13-2 | 397.63 | 100 MG
SDP
Catalog No. 5000388374
Encompass_Preferred
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute.
  • Can activate a conserved neural fasting response, thereby exerting an appetite-stimulating effect in rodents.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.
Catalog No. 50-003-88374 Supplier Medchemexpress LLC Supplier No. HYN0614R100MG
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute.
  • Can activate a conserved neural fasting response, thereby exerting an appetite-stimulating effect in rodents.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.