Learn More
Medchemexpress LLC Sucralose | 56038-13-2 | 397.63 | 50 MG
SDP
Catalog No. 5000388585
Encompass_Preferred
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute (E955; Trichlorosucrose). This compound can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Sucralose.
  • Intended for research and analytical applications.
  • Non-nutritive artificial sweetener and sugar substitute.
  • Activates a conserved neural fasting response, stimulating appetite in rodents.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Purity: 99.72%.
Catalog No. 50-003-88585 Supplier Medchemexpress LLC Supplier No. HYN0614R50MG
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute (E955; Trichlorosucrose). This compound can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Sucralose.
  • Intended for research and analytical applications.
  • Non-nutritive artificial sweetener and sugar substitute.
  • Activates a conserved neural fasting response, stimulating appetite in rodents.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Purity: 99.72%.