Learn More
Medchemexpress LLC Sucralose | 56038-13-2 | 397.63 | 500 MG
SDP
Catalog No. 5000388798
Encompass_Preferred
Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, which in turn exerts an appetite-stimulating effect in rodents. Its ability to activate a neural fasting response and stimulate appetite in rodents could be beneficial for specific research applications.
  • Non-nutritive artificial sweetener and sugar substitute
  • Activates a conserved neural fasting response
  • Stimulates appetite in rodents
Catalog No. 50-003-88798 Supplier Medchemexpress LLC Supplier No. HYN0614500MG
Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, which in turn exerts an appetite-stimulating effect in rodents. Its ability to activate a neural fasting response and stimulate appetite in rodents could be beneficial for specific research applications.
  • Non-nutritive artificial sweetener and sugar substitute
  • Activates a conserved neural fasting response
  • Stimulates appetite in rodents