Learn More
Cayman Chemical Sucralose-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 1459161-55-7, 1 mg
SDP
Catalog No. 502986250
Encompass_Preferred
An internal standard for the quantification of sucralose by GC- and LC-MS
Catalog No. 50-298-6250 Supplier Cayman Chemical Supplier No. 221761 MG
An internal standard for the quantification of sucralose by GC- and LC-MS