Learn More
Medchemexpress LLC Sucralose (E955; Trichlorosucrose) | 56038-13-2 | 397.63 | 1 ML
SDP
Catalog No. 5000388631
Encompass_Preferred
Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. This product is intended for research use only.
  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Activates neural fasting response
  • Appetite-stimulating effect in rodents
Catalog No. 50-003-88631 Supplier Medchemexpress LLC Supplier No. HYN061410MM/1ML
Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. This product is intended for research use only.
  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Activates neural fasting response
  • Appetite-stimulating effect in rodents