Learn More
Medchemexpress LLC Sucralose (Standard) | 56038-13-2 | 397.63 | 25 MG
SDP
Catalog No. 5000389554
Encompass_Preferred
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute that can activate a conserved neural fasting response, exerting an appetite-stimulating effect in rodents.
  • Serves as an analytical standard
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS
  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Can stimulate appetite in rodents by activating a neural fasting response
Catalog No. 50-003-89554 Supplier Medchemexpress LLC Supplier No. 25MGHYN0614R
Sucralose (Standard) is the analytical standard of Sucralose, intended for research and analytical applications. It is a non-nutritive artificial sweetener and sugar substitute that can activate a conserved neural fasting response, exerting an appetite-stimulating effect in rodents.
  • Serves as an analytical standard
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS
  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Can stimulate appetite in rodents by activating a neural fasting response