Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucralose, Thermo Scientific™

Catalog No. AC470090010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC470090010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC470090010 Supplier Thermo Scientific Chemicals Supplier No. 470090010
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector SUCRALOSE
Chemical Name or Material Sucralose
CAS 56038-13-2
Melting Point 130°C
Color White
Infrared Spectrum Conforms
Packaging Plastic bottle
Molecular Formula C12 H19 Cl3 O8
Quantity 1 kg
MDL Number 03648615
Merck Index XIII 8965
Specific Rotation Condition (20°C, 589 nm) (c=1% aq. soln.)
Specific Rotation 84° to 88°
Synonym Trichlorosucrose
Molecular Weight (g/mol) 397.63
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.