Learn More
Research Products International Corp Sucrose, 1 Kilogram
SDP
Supplier:  Research Products International Corp S240601000.0
Encompass_Preferred
Ultra-high purtiy, double crystallized to remove any remaining inorganic and organic contamination. Produces a clear colorless solution. Used for cultivating protoplasts and "in vitro" multiplication of plant tissue.
Catalog No. 50-213-671