Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose, 57-50-1, MFCD00006626, 1kg
Catalog No. 501657044
Encompass
Empirical Formula (Hill Notation) C12H22O11, ≥99.5% (GC), Molecular Weight 342.30, mp 185-187 °C (lit.), for molecular biology, Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar
Catalog No. 50-165-7044 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S03891KG
Add to Cart
Add to Cart
Empirical Formula (Hill Notation) C12H22O11, ≥99.5% (GC), Molecular Weight 342.30, mp 185-187 °C (lit.), for molecular biology, Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar