Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose, 57-50-1, MFCD00006626, 500g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S0389500G
Encompass
Empirical Formula (Hill Notation) C12H22O11, ≥99.5% (GC), Molecular Weight 342.30, mp 185-187 °C (lit.), for molecular biology, Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar
Catalog No. 50-165-7045