Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose, 57-50-1, MFCD00006626, 500 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S1888500G
Encompass
Empirical Formula (Hill Notation): C12H22O11, Molecular Weight: 342.30, Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar. BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC). Use to create sucrose gradients for purification of viruses and proteins.
Catalog No. NC0619477