Learn More
IBI SCIENTIFIC 3P Sucrose | 57-50-1 | MFCD00006626 | 5 kg
Supplier:  IBI SCIENTIFIC 3P IB37165
Encompass_Preferred
SUCROSE 5KG Sucrose is used in rate-zonal centrifugation to separate molecules, based on size and conformation.
Catalog No. NC0753491