Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose, 57-50-1, MFCD00006626, 5 KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S79035KG
Encompass
Empirical Formula (Hill Notation): C12H22O11, Molecular Weight: 342.30, BioXtra, ≥99.5% (GC), Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar. Sucrose may be used to prepare density gradients for cell/organelle separation. The refractive index of the solution will be directly related to the density. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media, and in various enzymatic assays.
Catalog No. NC0277937