Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK8360212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-8360-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-8360-212 Supplier Avantor Supplier No. 836006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 57-50-1
Packaging Poly
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 2.5 kg
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 5988
ChEBI CHEBI:17992
Formula Weight 342.3
Grade ACS
Physical Form Crystalline
Chemical Name or Material Sucrose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.