Learn More
Research Products International Corp Sucrose, ACS Grade, 10 KG
SDP
Catalog No. 501977407
Encompass_Preferred
Ultra-high purtiy, double crystallized to remove any remaining inorganic and organic contamination. Produces a clear colorless solution. Used for cultivating protoplasts and "in vitro" multiplication of plant tissue.
Catalog No. 50-197-7407 Supplier Research Products International Corp Supplier No. S2406510000.0
Ultra-high purtiy, double crystallized to remove any remaining inorganic and organic contamination. Produces a clear colorless solution. Used for cultivating protoplasts and "in vitro" multiplication of plant tissue.