Learn More
GFS Chemicals Inc SUCROSE, REAGENT (ACS) | 500 G | GFS CHEMICALS, INC. Item #: 70951 | Bulk #: 1044
SDP
Catalog No. NC1935980
Encompass_Preferred
SUCROSE, REAGENT (ACS) | Chemical Formula : C!1!2H!2!2O!1!1 | CAS # : 57-50-1 | Specifications : Chloride 0.005 % , Heavy metals (as Pb) 0.0005 % , Insoluble 0.005 % , Invert sugar (%) 0.05 % , Iron 0.0005 % , Loss on drying @ 105 C 0.03 % , Residue after ignition 0.01 % , Specific rotation 66.3 - 66.8, Sulfate and sulfite (%) 0.005 % , Titrable acid 0.0008 meq/g
Catalog No. NC1935980 Supplier GFS Chemicals Inc Supplier No. 70951
SUCROSE, REAGENT (ACS) | Chemical Formula : C!1!2H!2!2O!1!1 | CAS # : 57-50-1 | Specifications : Chloride 0.005 % , Heavy metals (as Pb) 0.0005 % , Insoluble 0.005 % , Invert sugar (%) 0.05 % , Iron 0.0005 % , Loss on drying @ 105 C 0.03 % , Residue after ignition 0.01 % , Specific rotation 66.3 - 66.8, Sulfate and sulfite (%) 0.005 % , Titrable acid 0.0008 meq/g