Learn More
Crescent Chemical Co Inc SUCROSE ANALYTICAL GRADE 500G
SDP
Supplier:  Crescent Chemical Co Inc SE35579.02
Encompass_Preferred
Sucrose analytical grade Free of DNA, RNA, DNase and RNase. Special grade for biochemistry and density gradient centrifugation. Tested for use in tissue culture. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35579.02 /
Catalog No. 50-248-165