Learn More
Medchemexpress LLC Sucrose-d14 | 99.0% | 5 MG
SDP
Catalog No. 5000413027
Encompass_Preferred
Sucrose-d14 is the deuterium labeled Sucrose. Sucrose (D-(+)-Saccharose) is a disaccharide composed of two monosaccharides, glucose and fructose. It can be applied in animal models for metabolic disease, obesity, and diet preference studies.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13027 Supplier Medchemexpress LLC Supplier No. HYB1779S35MG
Sucrose-d14 is the deuterium labeled Sucrose. Sucrose (D-(+)-Saccharose) is a disaccharide composed of two monosaccharides, glucose and fructose. It can be applied in animal models for metabolic disease, obesity, and diet preference studies.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.