Learn More
Medchemexpress LLC Sucrose-d14 (D-(+)-saccharose-d14) | 57-50-1 | 99.9% | 356.38 | C12H8D14O11 | 10 MG
SDP
Catalog No. 5000297283
Encompass_Preferred
Sucrose-d14 is the deuterium-labeled form of sucrose (D-(+)-saccharose), supplied as a high-purity solid for use in isotope-tracing, metabolic studies, and analytical workflows. It is intended for research use and provided in small quantities for precise experimental dosing.
  • High deuterium labeling for isotope-tracing experiments.
  • Chemical purity 99.9% reported by the manufacturer.
  • Supplied as a solid powder suitable for dissolution in common analytical solvents.
  • Available in small quantities for analytical and preparative studies.
  • Store powder at -20°C for long-term stability.
Catalog No. 50-002-97283 Supplier Medchemexpress LLC Supplier No. HYB1779S310MG
Sucrose-d14 is the deuterium-labeled form of sucrose (D-(+)-saccharose), supplied as a high-purity solid for use in isotope-tracing, metabolic studies, and analytical workflows. It is intended for research use and provided in small quantities for precise experimental dosing.
  • High deuterium labeling for isotope-tracing experiments.
  • Chemical purity 99.9% reported by the manufacturer.
  • Supplied as a solid powder suitable for dissolution in common analytical solvents.
  • Available in small quantities for analytical and preparative studies.
  • Store powder at -20°C for long-term stability.