Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sucrose diacetate hexaisobutyrate

Catalog No. AAJ66195A1
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66195A1 1 kg
AAJ6619530 250 g
AAJ6619536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66195A1 Supplier Thermo Scientific Chemicals Supplier No. J66195A1
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 27216-37-1 | C40H62O19

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27216-37-1
Molecular Formula C40H62O19
MDL Number MFCD00041859
Synonym SAIB

Specifications

Melting Point -7°C
Color Colorless to Yellow
Density 1.146
Boiling Point 288°C
Refractive Index 1.457
Quantity 1 kg
Optical Rotation +49° (c=1 in Chloroform)
Formula Weight 846.91
Physical Form Liquid
Chemical Name or Material Sucrose diacetate hexaisobutyrate
EINECSNumber 204-771-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.