Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose Fatty Acid Ester, TCI America™
SDP

Catalog No. S011225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011225G 25 g
S0112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S011225G Supplier TCI America Supplier No. S011225G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00148531
Synonym Fatty Acid Sucrose Ester, Saccharose Fatty Acid Ester

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Sucrose Fatty Acid Ester
EINECSNumber (8)-0067
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.