Learn More
Sigma Aldrich Fine Chemicals Biosciences Sucrose Grade I, suitable for plant cell culture | 57-50-1 | MFCD00006626 | 10KG
Catalog No. 501882421
Encompass
Sucrose Grade I, suitable for plant cell culture | Purity: >=99.5% (GC) | Mol Wt: 342.3 | 57-50-1 | MFCD00006626 | 10KG
Catalog No. 50-188-2421 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S539010KG
Sucrose Grade I, suitable for plant cell culture | Purity: >=99.5% (GC) | Mol Wt: 342.3 | 57-50-1 | MFCD00006626 | 10KG