Learn More
Medchemexpress LLC Sucrose hexasulfate, potassium salt | 386229-70-5 | C12H16K6O29S6 | 10MG
SDP
Catalog No. 5000259149
Encompass_Preferred
Sucrose hexasulfate (potassium salt) is a highly sulfated disaccharide used as a biochemical reagent and reference standard in glycobiology and analytical assays. Supplied in small, weighed quantities for laboratory research, it is used to study sulfated carbohydrate interactions, enzymatic processes, and as a standard for method development and characterization.
  • Highly sulfated disaccharide suitable for glycobiology research.
  • Provided as the potassium salt for stability and solubility.
  • Suitable as a reference standard in analytical and enzymatic assays.
  • Supplied in milligram-scale quantities for laboratory use.
  • Characterized by CAS 386229-70-5 and documented molecular weight (~1051 Da).
Catalog No. 50-002-59149 Supplier Medchemexpress LLC Supplier No. HY145501A10MG
Sucrose hexasulfate (potassium salt) is a highly sulfated disaccharide used as a biochemical reagent and reference standard in glycobiology and analytical assays. Supplied in small, weighed quantities for laboratory research, it is used to study sulfated carbohydrate interactions, enzymatic processes, and as a standard for method development and characterization.
  • Highly sulfated disaccharide suitable for glycobiology research.
  • Provided as the potassium salt for stability and solubility.
  • Suitable as a reference standard in analytical and enzymatic assays.
  • Supplied in milligram-scale quantities for laboratory use.
  • Characterized by CAS 386229-70-5 and documented molecular weight (~1051 Da).