Learn More
Cayman Chemical Sucrose hexasulfATpotasIu 10mg
SDP
Catalog No. 502966782
Encompass_Preferred
A polysulfated disaccharide that is used as a reference standard for sucralfate
Catalog No. 50-296-6782 Supplier Cayman Chemical Supplier No. 1823910 MG
A polysulfated disaccharide that is used as a reference standard for sucralfate