Learn More
Medchemexpress LLC Sucrose, monolaurate | 25339-99-5 | 95.0% | C24H46O13 | 1 G
SDP
Catalog No. 5000353666
Encompass_Preferred
Sucrose, monolaurate is a biochemical assay reagent intended exclusively for research use. It is supplied as a white to yellow solid, with specific storage requirements to maintain its stability, and is soluble in both water and DMSO.
  • Biochemical assay reagent
  • For research use only
  • White to yellow solid appearance
  • Soluble in water and DMSO
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C
Catalog No. 50-003-53666 Supplier Medchemexpress LLC Supplier No. HY1416331G
Sucrose, monolaurate is a biochemical assay reagent intended exclusively for research use. It is supplied as a white to yellow solid, with specific storage requirements to maintain its stability, and is soluble in both water and DMSO.
  • Biochemical assay reagent
  • For research use only
  • White to yellow solid appearance
  • Soluble in water and DMSO
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C