Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sucrose, monolaurate | 25339-99-5 | 97.0% | C24H46O13 | 100 MG
SDP

Catalog No. 5000356938
Encompass_Preferred

Sucrose, monolaurate is a biochemical assay reagent intended for research use only. It appears as a solid with a color ranging from white to yellow and has a molecular weight of 542.61.

  • Biochemical assay reagent
  • Solid appearance, white to yellow color
  • Molecular weight of 542.61
  • Soluble in H2O and DMSO (requires ultrasonic assistance)
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-56938 Supplier Medchemexpress LLC Supplier No. HY141633100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucrose, monolaurate is a biochemical assay reagent intended for research use only. It appears as a solid with a color ranging from white to yellow and has a molecular weight of 542.61.

  • Biochemical assay reagent
  • Solid appearance, white to yellow color
  • Molecular weight of 542.61
  • Soluble in H2O and DMSO (requires ultrasonic assistance)
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.