Learn More
Medchemexpress LLC Sucrose, monolaurate | 25339-99-5 | MFCD00077947 | 97.0% | C24H46O13 | 500 MG
SDP
Catalog No. 5000353704
Encompass_Preferred
Sucrose monolaurate is a non-ionic detergent and a carbohydrate fatty acid monoester. This biochemical assay reagent is supplied as a high-quality powder, primarily intended for research use. It is widely applied as a molecular tool in biochemical, chemical, and immunological studies.
  • Solubilizes membrane proteins
  • Acts as an antimicrobial agent
  • Used in lysis buffer for SPT activity determination
  • Improves disinfectant efficacy against E. coli O157:H7
  • Investigated for antibacterial properties against pathogenic bacteria
  • Exhibits inhibitory effects on Listeria monocytogenes and Staphylococcus aureus
Catalog No. 50-003-53704 Supplier Medchemexpress LLC Supplier No. HY141633500MG
Sucrose monolaurate is a non-ionic detergent and a carbohydrate fatty acid monoester. This biochemical assay reagent is supplied as a high-quality powder, primarily intended for research use. It is widely applied as a molecular tool in biochemical, chemical, and immunological studies.
  • Solubilizes membrane proteins
  • Acts as an antimicrobial agent
  • Used in lysis buffer for SPT activity determination
  • Improves disinfectant efficacy against E. coli O157:H7
  • Investigated for antibacterial properties against pathogenic bacteria
  • Exhibits inhibitory effects on Listeria monocytogenes and Staphylococcus aureus