Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sucrose octaacetate | 126-14-7 | 98.0% | 1 ML
SDP

Catalog No. 5000232373
Encompass_Preferred

Sucrose octaacetate is an acetylated derivative of sucrose (CAS 126-14-7) provided as a 10 mM solution in DMSO (1 mL). It is used as a bitter-tasting surrogate, food additive, adhesive/plasticizer, deterrent in formulations, and as a template for nanomaterial synthesis.

  • Provided as a 10 mM solution in DMSO for ready-to-use assays and formulations.
  • Acts as an intensely bitter compound suitable for sensory and deterrent studies.
  • Applicable as a food additive, adhesive, and plasticizer in formulation work.
  • Can be used as a template for synthesizing polyaniline nanofibers.
  • Supplied with CAS 126-14-7 and molecular formula C28H38O19 for identity confirmation.

Catalog No. 50-002-32373 Supplier Medchemexpress LLC Supplier No. HY11930910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucrose octaacetate is an acetylated derivative of sucrose (CAS 126-14-7) provided as a 10 mM solution in DMSO (1 mL). It is used as a bitter-tasting surrogate, food additive, adhesive/plasticizer, deterrent in formulations, and as a template for nanomaterial synthesis.

  • Provided as a 10 mM solution in DMSO for ready-to-use assays and formulations.
  • Acts as an intensely bitter compound suitable for sensory and deterrent studies.
  • Applicable as a food additive, adhesive, and plasticizer in formulation work.
  • Can be used as a template for synthesizing polyaniline nanofibers.
  • Supplied with CAS 126-14-7 and molecular formula C28H38O19 for identity confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.