Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sucrose octaacetate | 126-14-7 | 98.0% | 5 G
SDP

Catalog No. 5000234281
Encompass_Preferred

Sucrose octaacetate is an acetylated derivative of sucrose used as a bitter-tasting additive and biochemical reagent. Supplied as a white to off-white solid, it is used as a bittering agent, a template in polymer synthesis, and as a deterrent in formulations.

  • Purity 98.0%.
  • Molecular weight 678.59 g/mol.
  • Chemical formula C28H38O19.
  • CAS number 126-14-7.
  • Available in 5 g, 10 g, 50 g, and 10 mM (1 mL in DMSO) formats.
  • White to off-white solid with intensely bitter taste.

Catalog No. 50-002-34281 Supplier Medchemexpress LLC Supplier No. HY1193095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucrose octaacetate is an acetylated derivative of sucrose used as a bitter-tasting additive and biochemical reagent. Supplied as a white to off-white solid, it is used as a bittering agent, a template in polymer synthesis, and as a deterrent in formulations.

  • Purity 98.0%.
  • Molecular weight 678.59 g/mol.
  • Chemical formula C28H38O19.
  • CAS number 126-14-7.
  • Available in 5 g, 10 g, 50 g, and 10 mM (1 mL in DMSO) formats.
  • White to off-white solid with intensely bitter taste.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.