Learn More
Medchemexpress LLC Sucrose octasulfate (potassium) | 73264-44-5 | 98.0% | C12H14K8O35S8 | 250 MG
SDP
Catalog No. 5000352667
Encompass_Preferred
Sucrosofate potassium is a sucrose acid used to prepare liposomes for delivering Irinotecan, an inhibitor of topoisomerase Topo I. This compound is commonly employed in colon and rectal cancer research. Nanoliposome preparations loaded with Irinotecan are also known as PEP02 or MM-398.
  • Used in the preparation of liposomes.
  • Facilitates the delivery of Irinotecan.
  • Relevant for colon and rectal cancer research.
Catalog No. 50-003-52667 Supplier Medchemexpress LLC Supplier No. 250MGHY141436
Add to Cart
Add to Cart
Sucrosofate potassium is a sucrose acid used to prepare liposomes for delivering Irinotecan, an inhibitor of topoisomerase Topo I. This compound is commonly employed in colon and rectal cancer research. Nanoliposome preparations loaded with Irinotecan are also known as PEP02 or MM-398.
  • Used in the preparation of liposomes.
  • Facilitates the delivery of Irinotecan.
  • Relevant for colon and rectal cancer research.