Learn More
Cayman Chemical Sucrose octasulfate (potassium salt), Purity: ≥95%, CAS: 73264-44-5, 5 g
SDP
Catalog No. 502969933
Encompass_Preferred
An alkaline aluminum−sucrose complex that inhibits peptic hydrolysis and raises gastric pH, protecting esophageal epithelium against acid injury; inhibits thrombin activity (IC50 = 4.5 µM) and prevents FGF-2 binding to endothelial cells, inhibiting tumor growth in mouse melanoma and lung carcinoma models
Catalog No. 50-296-9933 Supplier Cayman Chemical Supplier No. 163825 G
An alkaline aluminum−sucrose complex that inhibits peptic hydrolysis and raises gastric pH, protecting esophageal epithelium against acid injury; inhibits thrombin activity (IC50 = 4.5 µM) and prevents FGF-2 binding to endothelial cells, inhibiting tumor growth in mouse melanoma and lung carcinoma models