Learn More
Crescent Chemical Co Inc Sucrose, 57-50-1, MFCD00006626, 1kg
SDP
Supplier:  Crescent Chemical Co Inc SE35580.02
Encompass_Preferred
Molecular Formula C12H22O11, Molecular Weight 342.295, Research Grade, For gradient centrifugation, bacteriology and electrophoresis. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-168