Learn More
Teknova Sucrose Selection Agar Plates. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387267
Encompass
1.0% Soytone, 0.5% Yeast Extract,1.5% Agar, 0.2% L-(+)-Arabinose, 6.0% Sucrose
Catalog No. 50-238-7267 Supplier Teknova Supplier No. S15264PK
1.0% Soytone, 0.5% Yeast Extract,1.5% Agar, 0.2% L-(+)-Arabinose, 6.0% Sucrose