Learn More
YSI 3P LifeScience 2780 Sucrose Standard, 5 g/L (250 mL)
Catalog No. 50200127
Encompass
5 g/L Sucrose standard solution. Default calibrator solution for the YSI 2700 and 2900 Series Analyzers.
Catalog No. 50-200-127 Supplier YSI 3P LifeScience Supplier No. 2780
5 g/L Sucrose standard solution. Default calibrator solution for the YSI 2700 and 2900 Series Analyzers.